Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ ZG16B Polyclonal Antibody, Invitrogen™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121493
                        
                            
                        
                        
                        
                            
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
ZG16B Polyclonal antibody specifically detects ZG16B in Human samples. It is validated for Western Blot.
            Specifications
| ZG16B | |
| Polyclonal | |
| Unconjugated | |
| ZG16B | |
| EECP; endocrine and exocrine protein; HRPE773; jacalin-like lectin domain containing 2; JCLN2; pancreatic adenocarcinoma upregulated factor; PAUF; PRO1567; UNQ773/PRO1567; ZG16B; Zymogen granule protein 16 homolog B; zymogen granule protein 16B | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 110259942, 124220 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96DA0 | |
| ZG16B | |
| Recombinant Zymogen Granule Protein 16 Homolog B (ZG16B), Gly17-Arg172 (Accession Q96DA0). | |
| 100 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction