Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZG16B Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

$249.00 - $566.00

Specifications

Antigen ZG16B
Concentration 0.5 mg/mL
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5121494
View Documents
Invitrogen™
PA5121494
20 μL
Each for $249.00
Only null left
Add to Cart
 
PIPA5121493
GREENER_CHOICE
View Documents
Invitrogen™
PA5121493
100 μL
Each for $566.00
Only null left
Add to Cart
 
Description

Description

ZG16B Polyclonal antibody specifically detects ZG16B in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZG16B
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Pig
Q96DA0
110259942, 124220
Recombinant Zymogen Granule Protein 16 Homolog B (ZG16B), Gly17-Arg172 (Accession Q96DA0).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
ZG16B
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
EECP; endocrine and exocrine protein; HRPE773; jacalin-like lectin domain containing 2; JCLN2; pancreatic adenocarcinoma upregulated factor; PAUF; PRO1567; UNQ773/PRO1567; ZG16B; Zymogen granule protein 16 homolog B; zymogen granule protein 16B
ZG16B
IgG
Antigen affinity chromatography, Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.