Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZGPAT Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB435975 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435975 0.025 mg
NBP241146 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435975 Supplier Novus Biologicals Supplier No. NBP2411460.025MG
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

ZGPAT Polyclonal specifically detects ZGPAT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZGPAT
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias GPATC6G patch domain-containing protein 6, GPATCH6Zinc finger and G patch domain-containing protein, KIAA1847Zinc finger CCCH domain-containing protein 9, MGC44880, ZC3H9FLJ14972, ZC3HDC9dJ583P15.3, zinc finger, CCCH-type with G patch domain, ZIPzinc finger CCCH-type with G patch domain-containing protein
Gene Symbols ZGPAT
Host Species Chicken
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ZGPAT. The immunogen is located within the last 50 amino acids of ZGPAT. Amino Acid Squence: CQEQRKADTHKKMTEF
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84619
Test Specificity Multiple isoforms of ZGPAT are known to exist.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.