Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZGPAT Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | ZGPAT |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ZGPAT Polyclonal specifically detects ZGPAT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ZGPAT | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
84619 | |
Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ZGPAT. The immunogen is located within the last 50 amino acids of ZGPAT. Amino Acid Squence: CQEQRKADTHKKMTEF | |
Primary | |
Multiple isoforms of ZGPAT are known to exist. |
1 mg/mL | |
Polyclonal | |
Chicken | |
Human | |
GPATC6G patch domain-containing protein 6, GPATCH6Zinc finger and G patch domain-containing protein, KIAA1847Zinc finger CCCH domain-containing protein 9, MGC44880, ZC3H9FLJ14972, ZC3HDC9dJ583P15.3, zinc finger, CCCH-type with G patch domain, ZIPzinc finger CCCH-type with G patch domain-containing protein | |
ZGPAT | |
IgY | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title