Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZGPAT Antibody, Novus Biologicals™
SDP

Catalog No. NBP15693720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15693720 20 μL
NBP156937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15693720 Supplier Novus Biologicals Supplier No. NBP15693720UL

Rabbit Polyclonal Antibody

ZGPAT Polyclonal specifically detects ZGPAT in Human samples. It is validated for Western Blot.

Specifications

Antigen ZGPAT
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q8N5A5
Gene Alias GPATC6G patch domain-containing protein 6, GPATCH6Zinc finger and G patch domain-containing protein, KIAA1847Zinc finger CCCH domain-containing protein 9, MGC44880, ZC3H9FLJ14972, ZC3HDC9dJ583P15.3, zinc finger, CCCH-type with G patch domain, ZIPzinc finger CCCH-type with G patch domain-containing protein
Gene Symbols ZGPAT
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ZGPAT(zinc finger, CCCH-type with G patch domain) The peptide sequence was selected from the middle region of ZGPAT. Peptide sequence LLREAVVEGDGILPPLRTEATESDSDSDGTGDSSYARVVGSDAVDSAQSS.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84619
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.