Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZGPAT Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZGPAT |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4019
![]() ![]() |
Proteintech Group Inc
232031AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Transcription repressor that specifically binds the 5′-GGAG[GA]A[GA]A-3′ consensus sequence. Represses transcription by recruiting the chromatin multiprotein complex NuRD to target promoters. Negatively regulates expression of EGFR, a gene involved in cell proliferation, survival and migration. Its ability to repress genes of the EGFR pathway suggest it may act as a tumor suppressor. Able to suppress breast carcinogenesis. Isoform 4: Antagonizes the transcription repression by isoform 1 by competing for the binding of the NuRD complex. Does not bind DNA.Specifications
ZGPAT | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8N5A5, Q8VDM1 | |
229007, 84619 | |
ZGPAT Fusion Protein Ag19620 | |
Primary | |
-20°C | |
ZGPAT |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GPATC6, GPATCH6, KIAA1847, ZC3H9, ZC3HDC9, ZGPAT, ZIP | |
ZGPAT | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title