Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zhangfei Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358004 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358004 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358004 Supplier Novus Biologicals Supplier No. NBP176539MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Zhangfei Polyclonal antibody specifically detects Zhangfei in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Zhangfei
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CREB/ATF bZIP transcription factor, FLJ94018, HCF-binding transcription factor Zhangfei, Host cell factor-binding transcription factor Zhangfei, SHP-interacting leucine zipper protein, SMILE, Zhangfei
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide near the amino terminus of human ZHANGFEI. The immunogen is located within amino acids 180 - 230 of ZHANGFEI. Amino Acid Squence: SGNDNNQAATKSPRKAA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 58487
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.