Learn More
Description
Specifications
Specifications
| Antigen | ZHX2 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.86mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Formulation | PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 |
| Gene Alias | Zona pellucida glycoprotein 2, zona pellucida glycoprotein 2 (sperm receptor), zona pellucida glycoprotein ZP2, Zona pellucida protein A, zona pellucida sperm-binding protein 2, Zp-2, ZPA |
| Gene Symbols | ZHX2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
