Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZHX3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10068219 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068219 Supplier Novus Biologicals Supplier No. NB10068219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZHX3 Polyclonal specifically detects ZHX3 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen ZHX3
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_055850.1
Gene Alias FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1
Gene Symbols ZHX3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human zinc fingers and homeoboxes 3 using the numbering given in entry NP_055850.1 (GeneID 23051).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23051
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.