Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIC3 Antibody (OTI1G7), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP274925IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274925IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274925IR Supplier Novus Biologicals Supplier No. NBP274925IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZIC3 Monoclonal specifically detects ZIC3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen ZIC3
Applications Western Blot
Classification Monoclonal
Clone OTI1G7
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias heterotaxy 1, HTXHTX1, Zic family member 3 (odd-paired homolog, Drosophila), Zinc finger protein 203, zinc finger protein ZIC 3, ZNF203Zinc finger protein of the cerebellum 3
Gene Symbols ZIC3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 242-467 of human ZIC3 (NP_003404) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7547.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.