Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika Virus (H/PF/2013) Envelope Antibody, Novus Biologicals™
SDP

Catalog No. NB176893 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176893 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176893 Supplier Novus Biologicals Supplier No. NBP313206
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Zika Virus (H/PF/2013) Envelope Polyclonal antibody specifically detects Zika Virus (H/PF/2013) Envelope in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence, ELISA

Specifications

Antigen Zika Virus (H/PF/2013) Envelope
Applications Western Blot, ELISA, Immunofluorescence, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:1000-1:10000, Immunofluorescence, Sandwich ELISA
Formulation PBS, 20% Glycerol (pH7)
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Zika Virus (H/PF/2013) Envelope (Zika virus (strain H/PF/2013)). The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.