Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika virus (H/PF/2013) NS1 Antibody (GT5212), Novus Biologicals™
SDP

Catalog No. NB177276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177276 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177276 Supplier Novus Biologicals Supplier No. NBP313589
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zika virus (H/PF/2013) NS1 Monoclonal antibody specifically detects Zika virus (H/PF/2013) NS1 in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence, ELISA

Specifications

Antigen Zika virus (H/PF/2013) NS1
Applications Western Blot, ELISA, Immunofluorescence, ELISA
Classification Monoclonal
Clone GT5212
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:1000-1:10000, Immunofluorescence, Sandwich ELISA
Formulation PBS
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Zika virus (H/PF/2013) NS1 (Zika virus (strain H/PF/2013)). The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.