Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika Virus IgG Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB163157 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163157 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163157 Supplier Novus Biologicals Supplier No. NBP261131AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Zika Virus IgG Polyclonal antibody specifically detects Zika Virus IgG in Virus samples. It is validated for ELISA

Specifications

Antigen Zika Virus IgG
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Zika Capsid antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Capsid. Amino Acid Squence: RIINARKEKKRRGADTS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.