Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika Virus IgG Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB413796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB413796 0.025 mg
NBP261132 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB413796 Supplier Novus Biologicals Supplier No. NBP2611320.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Zika Virus IgG Polyclonal specifically detects Zika Virus IgG in Viral samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Zika Virus IgG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Host Species Rabbit
Immunogen Zika Membrane antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Membrane. Amino Acid Squence: LPSHSTRKLQTRSQTWLE
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Zika
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.