Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus IgG Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Zika Virus IgG |
---|---|
Concentration | 1 mg/mL |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Zika Virus IgG Polyclonal specifically detects Zika Virus IgG in Viral samples. It is validated for Western Blot, ELISA.Specifications
Zika Virus IgG | |
Polyclonal | |
Rabbit | |
PBS with 0.02% Sodium Azide | |
IgG | |
Affinity Purified |
1 mg/mL | |
Unconjugated | |
Virus | |
Zika Membrane antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Membrane. Amino Acid Squence: LPSHSTRKLQTRSQTWLE | |
Primary | |
Zika |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title