Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika Virus (SPH2015) Envelope Antibody (033)
SDP

Catalog No. NB022141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022141 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB022141 Supplier Novus Biologicals Supplier No. NBP290343

Rabbit Monoclonal Antibody

Zika Virus (SPH2015) Envelope Monoclonal specifically detects Zika Virus (SPH2015) Envelope in Virus samples. It is validated for Sandwich ELISA Detection.

Specifications

Antigen Zika Virus (SPH2015) Envelope
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 33
Conjugate Unconjugated
Dilution Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Zika Virus (SPH2015) Envelope (ALU33341.1; Val593-Lys699).
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E/Envelope protein (Domain III)
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.