Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika Virus (SPH2015) Envelope Antibody (35), HRP , Novus Biologicals™
SDP

Catalog No. NB225341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB225341 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB225341 Supplier Novus Biologicals Supplier No. NBP305856H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zika Virus (SPH2015) Envelope Monoclonal antibody specifically detects Zika Virus (SPH2015) Envelope in Virus samples. It is validated for ELISA, ELISA

Specifications

Antigen Zika Virus (SPH2015) Envelope
Applications ELISA
Classification Monoclonal
Clone 35
Conjugate HRP
Formulation PBS
Gene Alias Zika Envelope
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika virus (strain Zika SPH2015) Envelope Protein (ZIKV-E) (ALU33341.1; Val593-Lys699).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.