Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus VLP Mouse anti-Virus, Clone: IG3-1-A12-G9, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP3148500.5MG
This item is not returnable.
View return policy
Description
Zika Virus VLP Monoclonal antibody specifically detects Zika Virus VLP in Virus samples. It is validated for Western Blot, ELISASpecifications
Zika Virus VLP | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA | |
polyprotein, partial | |
Recombinant Zika virus VLPs (comprising envelope, pre-membrane and membrane proteins), produced in HEK293 cells | |
0.5 mg | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Purified |
Western Blot, ELISA | |
IG3-1-A12-G9 | |
Unconjugated | |
PBS pH 7.4 | |
Mouse | |
Protein G purified | |
RUO | |
Virus | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction