Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIM2 Antibody (OTI6D12), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274928B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274928B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274928B Supplier Novus Biologicals Supplier No. NBP274928B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZIM2 Monoclonal specifically detects ZIM2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZIM2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6D12
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias zinc finger imprinted 2, zinc finger, imprinted 2, ZNF656Zinc finger protein 656
Gene Symbols ZIM2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-150 and 428-527 of human ZIM2 (NP_056178) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 23619.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.