Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc, 99.999%, (trace metal basis), powder, 40 mesh
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis, Reduction reagent
Specifications
Specifications
| Color | Gray |
| Melting Point | 420.0°C |
| Boiling Point | 907.0°C |
| Density | 7.14 |
| Quantity | 50 g |
| Packaging | Glass bottle |
| Form | Powder |
| Assay Percent Range | 100.00% |
| Linear Formula | Zn |
| Fieser | 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Very toxic to aquatic life with long lasting effects. In contact with water releases flammable gases which may ignite spontaneously. Catches fire spontaneously if exposed to air. |
| Precautionary Statement | GHS P Statement: Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction