Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc-alpha-2-glycoprotein Mouse anti-Human, Clone: 4B8A4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Zinc-alpha-2-glycoprotein |
---|---|
Clone | 4B8A4 |
Concentration | 0.98 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6612
![]() ![]() |
Proteintech Group Inc
661781IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
AZGP1 stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. It may bind polyunsaturated fatty acids.Specifications
Zinc-alpha-2-glycoprotein | |
0.98 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AZGP1, ZA2G, ZAG, Zinc alpha 2 glycoprotein, Zinc-alpha-2-glycoprotein, Zn alpha 2 glycoprotein, Zn alpha 2 GP, ZNGP1 | |
AZGP1 | |
IgG1 | |
Protein G | |
Antibody |
4B8A4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P25311 | |
563 | |
Zinc-alpha-2-glycoprotein Fusion Protein Ag5661 | |
Primary | |
-20°C | |
AZGP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title