Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc finger protein 639 Antibody (PCRP-ZNF639-2B2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB155440 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB155440 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB155440 Supplier Novus Biologicals Supplier No. NBP314131AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zinc finger protein 639 Monoclonal antibody specifically detects Zinc finger protein 639 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Zinc finger protein 639
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-ZNF639-2B2
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ANC_2H01, ZASC1ANC-2H01, zinc finger amplified in esophageal squamous cell carcinomas 1, zinc finger protein 639, Zinc finger protein ANC_2H01, Zinc finger protein ZASC1,6230400O18Rik
Host Species Mouse
Immunogen Recombinant human fragment (aa 406-485) of Zinc finger protein 639 (Uniprot: Q9UID6)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51193
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.