Learn More
Zinc nitrate hexahydrate, 99% (metals basis)
Description
Zinc nitrate hexahydrate is used in the synthesis of coordination polymers. It is utilized in the preparation of zinc oxide based nanowires. It is also used as a mordant in dyeing. It is involved in the preparation of a new microporous, organically templated zinc phosphate, which finds application as catalysts and other applications involving microporosity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∽36°C |
| Physical Form | Crystalline |
| Quantity | 10 kg |
| Assay Percent Range | 99% (metals basis) |
| Solubility Information | Soluble in water,alcoholSoluble in water and ethanol. |
| Formula Weight | 297.47 (189.38 Anhydrous) |
| Sensitivity | Hygroscopic |
| Density | 2.07 |
| Chemical Name or Material | Zinc nitrate hexahydrate |
Safety and Handling
| Hazard Category | H272-H302-H315-H319-H335 |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: II |
| EINECSNumber | 231-943-8 |
| RTECSNumber | ZH4775000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.