Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis)

Catalog No. p-7048654
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1113618 50 g
AA1113630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1113618 Supplier Thermo Scientific Chemicals Supplier No. 01113618
Only null left
Add to Cart

CAS: 10196-18-6 | H12N2O12Zn | 297.48 g/mol

Zinc nitrate hexahydrate is used as an analytical reagent, catalyst and as a mordant in dyeing. It finds application in electro galvanizing, rubber industry and in the water treatment industry. It is widely used as the precursor for the synthesis of zinc oxide. It is employed in the preparation of the steel phosphating agent. In the field of agriculture, it is used for nourishing and protecting fruit trees from acid rain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point ∼36°C
Color White
Physical Form Crystalline Aggregates
Quantity 50 g
Linear Formula Zn(NO3)2·6H2O
UN Number UN1514
Merck Index 14,10144
Solubility Information Soluble in water and alcohol.
IUPAC Name zinc;dinitrate;hexahydrate
Formula Weight 297.47 (189.38 Anhydrous)
Percent Purity 99.998%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Density 2.07 g/mL
Assay (metals basis)
Chemical Name or Material Zinc nitrate hexahydrate
Show More Show Less
Hazard Category H272-H302-H315-H319-H335
Hazard Statement GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: ZINC NITRATE
EINECSNumber 231-943-8
RTECSNumber ZH4775000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.