Learn More
Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Zinc oxide is incorporated in powders, ointments, and pastes. As Accelerator activator, pigment and reinforcing agent in rubber, ointments, pigment and mould-growth inhibitor in paints, uv absorber in plastics, ceramics, floor tile, glass, zinc salts, feed additive, dietary supplement, seed treatment cosmetics, photoconductor in office copying machines and in color photography, piezoelectric devices, artists' colorant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1975°C |
Physical Form | Tablets |
Quantity | 25 g |
UN Number | UN3077 |
Merck Index | 14,10147 |
Solubility Information | Insoluble in water. |
Formula Weight | 81.37 |
Percent Purity | 99.9% |
Odor | Odorless |
Assay | (metals basis) |
Show More |
Safety and Handling
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 215-222-5 |
RTECSNumber | ZH4810000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.