Alfa Aesar™ Zinc oxide, NanoArc™ ZN-2210, 40% in mineral spirits, colloidal dispersion with dispersant

Manufacturer: Alfa Aesar™ 4568322

Quantity 100g

 View more versions of this product

Catalog No. AA4568322

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1314-13-2
    Molecular Formula OZn
    Linear Formula ZnO
    Formula Weight 81.37
    MDL Number MFCD00011300
    UN Number UN1993
    Chemical Name or Material Zinc oxide, NanoArc™ ZN-2210, Colloidal dispersion with dispersant

    Specifications

    Physical Form Dry Powder (Typical)
    Formula Weight 81.37
    CAS 1314-13-2
    Odor Petroleum-like
    Molecular Formula OZn
    Linear Formula ZnO
    UN Number UN1993
    Color White
    Quantity 100g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Particle Size 20nm APS
    MDL Number MFCD00011300
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P273-P501a
    Avoid release to the environment.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 40% in mineral spirits
    Chemical Name or Material Zinc oxide, NanoArc™ ZN-2210, Colloidal dispersion with dispersant

    Description

    • Zinc oxide dispersed in mineral spirits suitable for incorporation in a wide variety of non-polar organic based coatings and formulations where high transparency is important

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P273-P501a
    Avoid release to the environment.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit