Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc p-toluenesulfonate hydrate, Thermo Scientific™

Catalog No. AC371080250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-05-4
Molecular Formula C14H14O6S2Zn
Molecular Weight (g/mol) 407.76
MDL Number MFCD00151013
InChI Key YISPIDBWTUCKKH-UHFFFAOYSA-L
Synonym zinc p-toluenesulfonate hydrate, acmc-20eh0l, p-toluenesulfonic acid zinc salt, zinc 2+ hydrate ditosylate, p-toluenesulfonic acid zinc salt hydrate, benzenesulfonic acid,4-methyl-, zinc salt, hydrate 2:1:?
PubChem CID 16212267
IUPAC Name zinc(2+) bis(4-methylbenzene-1-sulfonate)
SMILES [Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Packaging Glass bottle
Linear Formula (CH3C6H4SO3)2Zn·H2O
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 407.78
Chemical Name or Material Zinc p-toluenesulfonate hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.