Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc (Powder/Technical), Fisher Chemical

Catalog No. z463 Shop All Fisherbrand Products
Click to view available options
Quantity:
3 kg
Packaging:
Glass Bottle

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Blue
Melting Point 419°C
Boiling Point 908°C
Atomic Weight 65.37
Density 7.14g/cm³
Quantity 3 kg
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 1mmHg at 487°C
Form Solid
Formula Weight 65.41
Grade Technical
Chemical Name or Material Zinc
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.