Learn More
Zinc shot, 1-6mm (0.04-0.24in), Puratronic™, 99.9999% (metals basis)
Description
Polytetrafluoroethylene, 6-10 micron is used in specially designed coating applications on coil for bakeware.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Color | Gray |
| Appearance | Gray shot |
| Quantity | 250 g |
| Odor | Odorless |
| Form | Shot |
| Assay Percent Range | 99.9999% (Metals basis) |
| Formula Weight | 65.3 |
| Chemical Name or Material | Zinc, 1-6mm (0.04-0.24 in.) |
Safety and Handling
| EINECSNumber | 231-175-3 |
| RTECSNumber | ZG8600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.