Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc stearate

Catalog No. p-212524
Click to view available options
Quantity:
500 g
1 kg
5 kg
20 kg

CAS: 557-05-1 | C36H70O4Zn | 632.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-05-1
MDL Number MFCD00013031
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
Synonym zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat
PubChem CID 11178
IUPAC Name zinc;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2]

Specifications

Melting Point 118.0°C to 128.0°C
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Molecular Formula C36H70O4Zn
Linear Formula [CH3(CH2)16CO2]2Zn
Quantity 1 kg
Merck Index 15, 10358
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol,diethyl ether,,soluble in benzene,xylene,fats,oils
Molecular Weight (g/mol) 632.34
Formula Weight 632.34
Percent Purity 12.5 to 14.0% (ZnO)
Physical Form Powder
Chemical Name or Material Zinc stearate
Show More Show Less
EINECSNumber 209-151-9
RTECSNumber ZH5200000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.