Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Dihydrate, MACCO

Catalog No. 502985585
Click to view available options
Quantity:
12 kg
50 kg

Chemical Identifiers

CAS 10035-04-8
Molecular Weight (g/mol) 147.01 g/mol

Specifications

Physical Form Crystalline Powder
Quantity 50 kg
Percent Purity 99 to 100%
Grade USP
Packaging 50 kg poly drum with 2xPE liner
Chemical Name or Material Calcium chloride
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.