Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc tert-butoxide

Catalog No. AA4071803
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AA4071803

Supplier: Thermo Scientific Chemicals 04071803

Only null left
Add to Cart

CAS: 4278-43-7 | C8H18O2Zn | 211.61 g/mol

A sol-gel process has been developed for the chemical synthesis of ZnS at room temperature using zinc tert-butoxide and H 2 S as precursors in a toluene solution. The key step that enables this zinc-mediated epoxidation to be catalytic may be the equilibrium between zinc peroxide (R)-15 and zinc tert-butoxide (R)-16 in the presence of t-BuOOH. An additional important feature that renders the dual catalytic cycles feasible is the poor reactivity of zinc tert-butoxide toward the iridium-bound cationic intermediate, whereby the requisite deprotonation process is effectively mediated by zinc tert-butoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4278-43-7
Molecular Formula C8H18O2Zn
Molecular Weight (g/mol) 211.61
MDL Number MFCD00145545
InChI Key DVGVEVPHJQJMPP-UHFFFAOYSA-N
Synonym zinc tert-butoxide, di-tert-butoxyzinc, acmc-20aj87, zinc bis 2-methylpropan-2-olate, zinc 2+ bis 2-methylpropan-2-olate, zinc 2+ ion bis 2-methylpropan-2-olate
PubChem CID 14178434
IUPAC Name zinc;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].[Zn+2]

Specifications

Color White to Yellow
Odor Odorless
Linear Formula Zn[OC(CH3)3]2
Quantity 1 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 211.61
Physical Form Powder
Chemical Name or Material Zinc tert-butoxide
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.