Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc trifluoroacetate hydrate

Catalog No. p-7049692
Click to view available options
Quantity:
25 g
100 g

CAS: 207801-31-8 | C4F6O4Zn | 291.41 g/mol

Zinc trifluoroacetate hydrate is useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207801-31-8
Molecular Formula C4F6O4Zn
Molecular Weight (g/mol) 291.41
MDL Number MFCD00061595
InChI Key VCQWRGCXUWPSGY-UHFFFAOYSA-L
Synonym zinc trifluoroacetate hydrate, acmc-1ch94, zinc trifluoroacetate-water 1/2/1, zinc 2+ hydrate ditrifluoroacetate
PubChem CID 57375648
IUPAC Name zinc;2,2,2-trifluoroacetate;hydrate
SMILES [Zn++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Quantity 25 g
UN Number UN3077
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 291.40 (Anhydrous)
Physical Form Chunks and/or Powder
Chemical Name or Material Zinc trifluoroacetate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.