Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc trifluoromethanesulfonate, 98%

Catalog No. p-7046707
Click to view available options
Quantity:
5 g
25 g

CAS: 54010-75-2 | C2F6O6S2Zn | 363.51 g/mol

Zinc trifluoromethanesulfonate acts as a catalyst for the preparation of dithioketals. It is used as a Lewis acid catalyst in silylation reactions. It is also used as a catalyst for greener amine synthesis by reductive amination with hydrogen gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc trifluoromethanesulfonate acts as a catalyst for the preparation of dithioketals. It is used as a Lewis acid catalyst in silylation reactions. It is also used as a catalyst for greener amine synthesis by reductive amination with hydrogen gas.

Solubility
Soluble in water and acetonitrile. Slightly soluble in methanol. Insoluble in dichloromethane.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54010-75-2
Molecular Formula C2F6O6S2Zn
Molecular Weight (g/mol) 363.51
MDL Number MFCD00013229
InChI Key CITILBVTAYEWKR-UHFFFAOYSA-L
Synonym zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
PubChem CID 104671
SMILES [Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point >300°C
Linear Formula (CF3SO3)2Zn
Quantity 5 g
UN Number UN3261
Beilstein 4028195
Sensitivity Hygroscopic
Solubility Information Soluble in water and acetonitrile. Slightly soluble in methanol. Insoluble in dichloromethane.
IUPAC Name zinc(2+) ditrifluoromethanesulfonate
Formula Weight 363.51
Percent Purity 98%
Chemical Name or Material Zinc trifluoromethanesulfonate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 258-922-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.