Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIP4H/Tex11 Antibody, Novus Biologicals™
SDP

Catalog No. NB036258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036258 Supplier Novus Biologicals Supplier No. NBP288636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZIP4H/Tex11 Polyclonal specifically detects ZIP4H/Tex11 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZIP4H/Tex11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias testis expressed 11, testis expressed sequence 11, testis-expressed sequence 11 protein, TGC1, TSGA3
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of Human ZIP4H/Tex11
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56159
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.