Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) oxide, 99.7% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Catalog No. aa4152818
Click to view available options
Quantity:
50 g
250 g
1 kg

Zirconium(IV) oxide is used as an additive in the production of piezoelectric crystals, in high-frequency induction coils and in ceramic glazes. It finds application as solid electrolyte in electrochromic devices and precursor to the electroceramic lead zirconate titanate (PZT), as insulator in transistors, as a thermal barrier coating in jet and diesel engines and as diamond simulant in jewelry. It is used in oxygen sensors, fuel cell membranes and in orthopaedic implants of hip joints. It is coated on silica gel surface and found to be used in the extraction of chromium from aqueous solutions at a pH of about 3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Boiling Point ∼5000°C
Melting Point ∼2700°C
Physical Form Powder
Quantity 50 g
Merck Index 14,10180
Solubility Information Soluble in alcohols,hydrofluoric and hot sulphuric acid. Insoluble in water.
Formula Weight 123.22
Percent Purity 99.7%
Odor Odorless
Density 5.89 g/mL
Assay (metals basis excluding Hf), Hf <75ppm
Chemical Name or Material Zirconium(IV) oxide
Show More Show Less
EINECSNumber 215-227-2
RTECSNumber ZH8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.