Learn More
Zirconium(IV) oxide, 99+% (metals basis excluding Hf), HfO{2} 2%, Thermo Scientific Chemicals
Description
Zirconium(IV) oxide is used in the production of piezoelectric crystals and also used in the high frequency induction coils and ceramic glazes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | ca 5000°C |
| Melting Point | ∽2700°C |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | ≥99%, metal basis |
| Solubility Information | Insoluble in water. Soluble in sulfuric acid and hydrofluoric acid. |
| Formula Weight | 123.22 |
| Density | 5.89 |
| Chemical Name or Material | Zirconium(IV) Oxide |
Safety and Handling
| EINECSNumber | 215-227-2 |
| RTECSNumber | ZH8800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.