Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium silicate, Thermo Scientific Chemicals

Catalog No. AA3047736
Click to view available options
Quantity:
500 g
2 kg
10 kg

Catalog No. AA3047736

Supplier: Thermo Scientific Chemicals 03047736

Only null left
Add to Cart

Zirconium silicate is used for manufacturing refractory materials for applications where resistance to corrosion by alkali materials is required. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. It can be also present in some cements. Another use of zirconium silicate is as beads for milling and grinding. Thin films of zirconium silicate and hafnium silicate produced by chemical vapor deposition. Potential alternative gate dielectric to silica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-52-7
Molecular Formula O4SiZr
Molecular Weight (g/mol) 183.31
MDL Number MFCD00085353
InChI Key GFQYVLUOOAAOGM-UHFFFAOYSA-N
Synonym zirconium silicate, hyacinth, zircon, zircon zr sio4, zirconite, zircosil 15, standard sf 200, ultrox 500w, a-pax 45m, zirconium iv silicate
PubChem CID 61775
IUPAC Name zirconium(4+);silicate
SMILES [Zr+4].[O-][Si]([O-])([O-])[O-]

Specifications

Melting Point 2550°C
Color White
Physical Form Powder
Quantity 500 g
Merck Index 14,10181
Solubility Information Insoluble in water,acids,alkali and aqua regia.
Formula Weight 183.3
Odor Odorless
Density 4.56 g/mL
Chemical Name or Material Zirconium silicate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 233-252-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.