Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel(II) chloride hexahydrate, 98+%
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| pH | 4 to 6 (5% soln. at 20°C) |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 1 kg |
| Assay Percent Range | 98% min. (Complexometry) |
| Linear Formula | NiCl2·6H2O |
| Fieser | 14,351; 15,96; 17,86 |
| Merck Index | 15,659 |
| Solubility Information | Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol |
| Formula Weight | 237.71 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Toxic if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. Very toxic to aquatic life with long lasting effects.<br |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Avoid relea |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction