Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZJ 43

Catalog No. p-7112878
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Glutamate carboxypeptidase II and III (NAALADase, NAAG peptidase) inhibitor

Potent inhibitor of glutamate carboxypeptidase II and III (GCP II and III/NAAG peptidase/NAALADase) (Ki values are 0.8 and 23nM respectively) that inhibits the hydrolysis of NAAG (IC50 = 2.4nM) . Does not directly interact with NMDA or metabotropic glutamate receptors. Reduces neuronal degeneration in a rat model of? traumatic brain injury (TBI) and reduces locomotor activity in the PCP-model of schizophrenia.

Specifications

Chemical Name or Material N-[[[(1S)-1-Carboxy-3-methylbutyl]amino]carbonyl]-L-glutamic acid
CAS 723331-20-2
Quantity 1 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C12H20N2O7
Formula Weight Observed MW: 304.3

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.