Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZK 200775
GREENER_CHOICE

Catalog No. 234510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-451-0 10 mg
23-455-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-451-0 Supplier Tocris Bioscience™ Supplier No. 2345/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Competitive non-NMDA iGluR antagonist

Competitive AMPA/kainate antagonist. In rat cortical membranes, displays high affinity for [3H]-AMPA (Ki = 120 nM) and [3H]-CNQX (Ki = 32 nM) binding sites and low affinity for kainate and NMDA channel-associated binding sites (IC50 values range from 2.5 to 11 μM). Inhibits currents induced by AMPA, kainate and NMDA with IC50 values of 21 nM, 27 nM, and > 1 μM respectively. Displays anxiolytic, anticonvulsant and muscle relaxant activity in vivo.

Chemical Identifiers

CAS 161605-73-8
Molecular Formula C14H15F3N3O6P

Specifications

Quantity 10 mg
Formula Weight 409.25
Purity >99%
Molecular Weight (g/mol) 409.258
Chemical Name or Material ZK 200775
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.