Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZM 241385
GREENER_CHOICE

Catalog No. 103610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-361-0 10 mg
10-365-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-361-0 Supplier Tocris Bioscience™ Supplier No. 1036/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, highly selective A2A antagonist

Potent and highly selective A2A adenosine antagonist, with a pA2 of 9.02 for A2A receptors in guinea pig cardiac vasculature and fold selectivities of 1000, 91 and 500,000 over A1, A2B and A3 sites respectively. Active in vivo.

Chemical Identifiers

CAS 139180-30-6
Molecular Formula C16H15N7O2

Specifications

Quantity 10 mg
Formula Weight 337.34
Purity >99%
Molecular Weight (g/mol) 337.343
Chemical Name or Material ZM 241385
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.