Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMAT4 Antibody, Novus Biologicals™
SDP

Catalog No. NB395594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395594 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395594 Supplier Novus Biologicals Supplier No. NBP24961925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZMAT4 Polyclonal antibody specifically detects ZMAT4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ZMAT4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias FLJ13842, zinc finger matrin-type protein 4, zinc finger, matrin type 4, zinc finger, matrin-type 4
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: QHYDGKKHKKNAARVALLEQLGTTLDMGELRGLRRNYRCTICSVSLNSIEQYH
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79698
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.