Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMPSTE24 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB339794 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339794 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339794 Supplier Novus Biologicals Supplier No. NB1002388PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZMPSTE24 Polyclonal specifically detects ZMPSTE24 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZMPSTE24
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias zona pellucida binding protein 2, zona pellucida-binding protein 2, ZPBP-like protein, ZPBPLMGC41930
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ZMPSTE24 protein sequence (between residues 400-475). [Swiss-Prot# O75844]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10269
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.