Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMYM3 Antibody (C.15200016), Novus Biologicals™
SDP

Catalog No. NBP259244 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259244 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259244 Supplier Novus Biologicals Supplier No. NBP259244
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZMYM3 Monoclonal specifically detects ZMYM3 in Human samples. It is validated for Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Protein Array, Chromatin Immunoprecipitation Sequencing.

Specifications

Antigen ZMYM3
Applications Immunocytochemistry, Immunofluorescence, Protein Array, ChIP Assay
Classification Monoclonal
Clone C.15200016
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:1000, Protein Array 1:100000, Chromatin Immunoprecipitation (ChIP) 1-2 ug, Chromatin Immunoprecipitation Sequencing 1 - 2 ug/ChIP
Formulation PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Gene Alias DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3
Gene Symbols ZMYM3
Host Species Mouse
Immunogen The exact immunogen is proprietary information.
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9203
Target Species Human
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.