Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMYM4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB341333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341333 Supplier Novus Biologicals Supplier No. NBP282054MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZMYM4 Polyclonal antibody specifically detects ZMYM4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ZMYM4
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CDIR, cell death inhibiting RNA, KIAA0425DKFZp686L0547, MYM, zinc finger MYM-type protein 4, Zinc finger protein 262DKFZp686B09210, zinc finger, MYM-type 4, ZNF198L3, ZNF262
Host Species Rabbit
Immunogen ZMYM4 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZMYM4. The immunogen is located within the last 50 amino acids of ZMYM4. Amino Acid Squence: EVHEELAKAKSEDSDVE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 9202
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.