Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF10 Antibody (CL9747), Novus Biologicals™
SDP

Catalog No. NB036535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036535 Supplier Novus Biologicals Supplier No. NBP288940
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF10 Monoclonal specifically detects ZNF10 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZNF10
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone CL9747
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol
Gene Alias KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids: RDQLDKYQENPERHLRQV
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7556
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.