Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF114 Antibody (OTI6F4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274931A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274931A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274931A30 Supplier Novus Biologicals Supplier No. NBP274931AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF114 Monoclonal specifically detects ZNF114 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF114
Applications Western Blot
Classification Monoclonal
Clone OTI6F4
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias MGC149700, MGC17986, zinc finger protein 114, zinc finger protein 114 (Y18)
Gene Symbols ZNF114
Host Species Mouse
Immunogen Full length recombinant protein of human ZNF114 (NP_705836) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 163071
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.