Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF131 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110947
Description
Immunogen sequence: GRKKLYECQV CNSVFNSWDQ FKDHLVIHTG DKPNHCTLCD LWFMQGNELR RHLSDAHNIS ERLVTEEVLS VETRVQTEPV TSMTIIEQVG KVHVLPLLQV QVDSAQVTVE QVHPDLLQD.
May be involved in transcriptional regulation. Plays a role during development and organogenesis as well as in the function of the adult central nervous system.
Specifications
ZNF131 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P52739 | |
ZNF131 | |
Recombinant Protein corresonding to Human ZNF131. Recombinant protein control fragment (Product #RP-105779). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
Unconjugated | |
ZNF131 | |
2610109I01Rik; AW558871; pHZ-10; ZBTB35; Zfp131; zinc finger and BTB domain containing 35; zinc finger protein 131; zinc finger protein 131 (clone pHZ-10); Zinc finger protein 131-like protein; ZNF131 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7690 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction