Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF131 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | ZNF131 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5110947
|
Invitrogen™
PA5110947 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111722
|
Invitrogen™
PA5111722 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: GRKKLYECQV CNSVFNSWDQ FKDHLVIHTG DKPNHCTLCD LWFMQGNELR RHLSDAHNIS ERLVTEEVLS VETRVQTEPV TSMTIIEQVG KVHVLPLLQV QVDSAQVTVE QVHPDLLQD.
May be involved in transcriptional regulation. Plays a role during development and organogenesis as well as in the function of the adult central nervous system.Specifications
ZNF131 | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P52739 | |
7690 | |
Recombinant Protein corresonding to Human ZNF131. Recombinant protein control fragment (Product #RP-105779). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
ZNF131 | |
2610109I01Rik; AW558871; pHZ-10; ZBTB35; Zfp131; zinc finger and BTB domain containing 35; zinc finger protein 131; zinc finger protein 131 (clone pHZ-10); Zinc finger protein 131-like protein; ZNF131 | |
ZNF131 | |
Primary | |
Antigen Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title